Product Details

SNP ID
rs16826967
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:145933847 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCTGGAGAATTCAAGGAAGCCC[G/A]TTTCAAGCACTAATAAAATTGTAGC
Phenotype
MIM: 612875 MIM: 605313
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GNRHR2 PubMed Links

Gene Details

Gene
GNRHR2
Gene Name
gonadotropin releasing hormone receptor 2 (pseudogene)
There are no transcripts associated with this gene.

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153713.2 3569 UTR 3 NP_714924.1
XM_017000316.1 3569 UTR 3 XP_016855805.1
Gene
RBM8A
Gene Name
RNA binding motif protein 8A
There are no transcripts associated with this gene.

View Full Product Details