Product Details

SNP ID
rs17052984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:122476786 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACATTTCTAAAACATTTCACTTT[C/T]CAAGGGCAGATGTATTGACCTTGAT
Phenotype
MIM: 606914 MIM: 614548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKIB PubMed Links

Gene Details

Gene
PKIB
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270393.1 Intron NP_001257322.1
NM_001270394.1 Intron NP_001257323.1
NM_001270395.1 Intron NP_001257324.1
NM_032471.5 Intron NP_115860.1
NM_181794.2 Intron NP_861459.1
NM_181795.2 Intron NP_861460.1
XM_011535930.2 Intron XP_011534232.1
XM_011535931.2 Intron XP_011534233.1
XM_011535932.2 Intron XP_011534234.1
XM_011535933.2 Intron XP_011534235.1
XM_011535934.2 Intron XP_011534236.1
XM_011535935.2 Intron XP_011534237.1
XM_011535936.1 Intron XP_011534238.1
XM_011535937.2 Intron XP_011534239.1
Gene
SERINC1
Gene Name
serine incorporator 1
There are no transcripts associated with this gene.

View Full Product Details