Product Details

SNP ID
rs17050639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9904294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTGGATTATCCTAAAATAGCAC[G/T]CTATGACTGATATTCACAAATACAA
Phenotype
MIM: 614995 MIM: 616012
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL17RE PubMed Links

Gene Details

Gene
IL17RE
Gene Name
interleukin 17 receptor E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193380.1 Intron NP_001180309.1
NM_153480.1 Intron NP_705613.1
NM_153481.1 Intron NP_705614.1
NM_153483.2 Intron NP_705616.2
XM_006712975.1 Intron XP_006713038.1
XM_006712976.1 Intron XP_006713039.1
XM_011533361.2 Intron XP_011531663.1
XM_011533362.2 Intron XP_011531664.1
XM_011533363.2 Intron XP_011531665.1
XM_011533364.2 Intron XP_011531666.1
Gene
JAGN1
Gene Name
jagunal homolog 1
There are no transcripts associated with this gene.

View Full Product Details