Product Details

SNP ID
rs35833816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74455814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATCTTGGCTTACTGCAACCTCC[A/G]CCTCCCGGGTTCAAACAATTCTCCT
Phenotype
MIM: 604433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNE3 PubMed Links

Gene Details

Gene
KCNE3
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005472.4 2167 UTR 3 NP_005463.1
XM_011544713.2 2167 Intron XP_011543015.1
XM_017017047.1 2167 UTR 3 XP_016872536.1
XM_017017048.1 2167 UTR 3 XP_016872537.1
XM_017017049.1 2167 UTR 3 XP_016872538.1
XM_017017050.1 2167 UTR 3 XP_016872539.1
XM_017017051.1 2167 UTR 3 XP_016872540.1
XM_017017052.1 2167 UTR 3 XP_016872541.1

View Full Product Details