Product Details

SNP ID
rs16857666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183488603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTGCCATGTCCTGCCTGAGTC[C/T]GAGGGTTCTTTGTCTCTGGGTTTAT
Phenotype
MIM: 614214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL6 PubMed Links

Gene Details

Gene
KLHL6
Gene Name
kelch like family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130446.2 4903 UTR 3 NP_569713.2
XM_011513273.2 4903 UTR 3 XP_011511575.1
XM_011513274.2 4903 Intron XP_011511576.1

View Full Product Details