Product Details

SNP ID
rs7853287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114787047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGATCATGGTCTCTGTATTTGCT[A/G]AGGAACACAGATTTTCAAAGCATTT
Phenotype
MIM: 604052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF15 PubMed Links

Gene Details

Gene
TNFSF15
Gene Name
tumor necrosis factor superfamily member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204344.1 4277 UTR 3 NP_001191273.1
NM_005118.3 4277 UTR 3 NP_005109.2

View Full Product Details