Product Details

SNP ID
rs16871363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104382017 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATTCTCCATTTGCCATTTACTAC[A/G]CATAGGAAACTGACAAGTTAATTAA
Phenotype
MIM: 613326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPYS PubMed Links

Gene Details

Gene
DPYS
Gene Name
dihydropyrimidinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001385.2 Intron NP_001376.1
XM_005250818.3 Intron XP_005250875.1
XM_006716518.3 Intron XP_006716581.1
XM_011516903.2 Intron XP_011515205.1
XM_017013167.1 Intron XP_016868656.1

View Full Product Details