Product Details

SNP ID
rs17227606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98732853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGGTAAAATTCTGTCTTGATG[C/T]ATTCAAAATGGTTGACATAATTAAT
Phenotype
MIM: 607815 MIM: 602233
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 7016 Intron NP_001190994.1
NM_001204066.1 7016 Intron NP_001190995.1
NM_001204067.1 7016 Intron NP_001190996.1
NM_001204068.1 7016 Intron NP_001190997.1
NM_001204069.1 7016 Intron NP_001190998.1
NM_001204070.1 7016 Intron NP_001190999.1
NM_001204079.1 7016 Intron NP_001191008.1
NM_001204080.1 7016 Intron NP_001191009.1
NM_001204081.1 7016 Intron NP_001191010.1
NM_020140.3 7016 Intron NP_064525.1
NM_152788.4 7016 Intron NP_690001.3
NM_181670.3 7016 Intron NP_858056.2
XM_005269028.4 7016 Intron XP_005269085.1
XM_005269029.4 7016 Intron XP_005269086.1
XM_005269032.3 7016 Intron XP_005269089.1
XM_006719504.3 7016 Intron XP_006719567.1
XM_006719505.3 7016 Intron XP_006719568.1
XM_006719506.3 7016 Intron XP_006719569.1
XM_006719507.3 7016 Intron XP_006719570.1
XM_006719508.3 7016 Intron XP_006719571.1
XM_006719509.3 7016 Intron XP_006719572.1
XM_006719510.3 7016 Intron XP_006719573.1
XM_006719512.3 7016 Intron XP_006719575.1
XM_006719513.3 7016 Intron XP_006719576.1
XM_006719514.3 7016 Intron XP_006719577.1
XM_011538571.2 7016 Intron XP_011536873.1
XM_017019651.1 7016 Intron XP_016875140.1
XM_017019652.1 7016 Intron XP_016875141.1
XM_017019653.1 7016 UTR 3 XP_016875142.1
XM_017019654.1 7016 Intron XP_016875143.1
XM_017019655.1 7016 Intron XP_016875144.1
XM_017019656.1 7016 Intron XP_016875145.1
XM_017019657.1 7016 Intron XP_016875146.1
XM_017019658.1 7016 Intron XP_016875147.1
XM_017019659.1 7016 Intron XP_016875148.1
XM_017019660.1 7016 Intron XP_016875149.1
XM_017019661.1 7016 Intron XP_016875150.1
XM_017019662.1 7016 Intron XP_016875151.1
XM_017019663.1 7016 Intron XP_016875152.1
XM_017019664.1 7016 Intron XP_016875153.1
XM_017019665.1 7016 Intron XP_016875154.1
Gene
APAF1
Gene Name
apoptotic peptidase activating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160.2 7016 UTR 3 NP_001151.1
NM_013229.2 7016 UTR 3 NP_037361.1
NM_181861.1 7016 UTR 3 NP_863651.1
NM_181868.1 7016 UTR 3 NP_863658.1
NM_181869.1 7016 UTR 3 NP_863659.1
XM_017019250.1 7016 Intron XP_016874739.1

View Full Product Details