Product Details

SNP ID
rs16883896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34287854 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAGTAATCCCAAGAATCCAGTTC[A/C]AACTCTACTGTCAACATGTGGGGCA
Phenotype
MIM: 609228
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC102723521 PubMed Links

Gene Details

Gene
LOC102723521
Gene Name
uncharacterized LOC102723521
There are no transcripts associated with this gene.

Gene
NUDT3
Gene Name
nudix hydrolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006703.3 1869 UTR 3 NP_006694.1
Gene
RPS10-NUDT3
Gene Name
RPS10-NUDT3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202470.2 1869 UTR 3 NP_001189399.1

View Full Product Details