Product Details

SNP ID
rs34883442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:56358938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAAACCTCCGCCAGATGCTGCA[C/G]ATGCTTCTCCTGCCAGGAGTGAGCA
Phenotype
MIM: 609177
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZWINT PubMed Links

Gene Details

Gene
ZWINT
Gene Name
ZW10 interacting kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005413.1 527 Missense Mutation CTG,GTG L164V NP_001005413.1
NM_007057.3 527 Missense Mutation CTG,GTG L164V NP_008988.2
NM_032997.2 527 Missense Mutation CTG,GTG L164V NP_127490.1
XM_017015605.1 527 Missense Mutation CTG,GTG L164V XP_016871094.1

View Full Product Details