Product Details

SNP ID
rs17168376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135450562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCCTAGAAACCCTGGTGAAGAGT[A/C]AACAGCTAGCACATTCCGAGTGGTT
Phenotype
MIM: 609710 MIM: 612663
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCANP1 PubMed Links

Gene Details

Gene
DCANP1
Gene Name
dendritic cell associated nuclear protein
There are no transcripts associated with this gene.

Gene
TIFAB
Gene Name
TIFA inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099221.1 Intron NP_001092691.1

View Full Product Details