Product Details

SNP ID
rs17500814
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:225488880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCAAATGGCACGGAAGCGGCACC[C/T]GGTCTGCCAAGACATTGCTGTTAAA
Phenotype
MIM: 609061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENAH PubMed Links

Gene Details

Gene
ENAH
Gene Name
enabled homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008493.1 10890 UTR 3 NP_001008493.1
NM_018212.4 10890 UTR 3 NP_060682.2
XM_005273187.2 10890 UTR 3 XP_005273244.1
XM_011544229.2 10890 Intron XP_011542531.2
XM_017001746.1 10890 UTR 3 XP_016857235.1
XM_017001747.1 10890 Intron XP_016857236.1
XM_017001748.1 10890 Intron XP_016857237.1
XM_017001749.1 10890 Intron XP_016857238.1
XM_017001750.1 10890 Intron XP_016857239.1
XM_017001751.1 10890 UTR 3 XP_016857240.1
XM_017001752.1 10890 Intron XP_016857241.1
XM_017001753.1 10890 UTR 3 XP_016857242.1
XM_017001754.1 10890 UTR 3 XP_016857243.1
XM_017001755.1 10890 UTR 3 XP_016857244.1
XM_017001756.1 10890 UTR 3 XP_016857245.1
XM_017001757.1 10890 UTR 3 XP_016857246.1

View Full Product Details