Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001008493.1 | 8087 | UTR 3 | NP_001008493.1 | ||
NM_018212.4 | 8087 | UTR 3 | NP_060682.2 | ||
XM_005273187.2 | 8087 | UTR 3 | XP_005273244.1 | ||
XM_011544229.2 | 8087 | Intron | XP_011542531.2 | ||
XM_017001746.1 | 8087 | UTR 3 | XP_016857235.1 | ||
XM_017001747.1 | 8087 | Intron | XP_016857236.1 | ||
XM_017001748.1 | 8087 | Intron | XP_016857237.1 | ||
XM_017001749.1 | 8087 | Intron | XP_016857238.1 | ||
XM_017001750.1 | 8087 | Intron | XP_016857239.1 | ||
XM_017001751.1 | 8087 | UTR 3 | XP_016857240.1 | ||
XM_017001752.1 | 8087 | Intron | XP_016857241.1 | ||
XM_017001753.1 | 8087 | UTR 3 | XP_016857242.1 | ||
XM_017001754.1 | 8087 | UTR 3 | XP_016857243.1 | ||
XM_017001755.1 | 8087 | UTR 3 | XP_016857244.1 | ||
XM_017001756.1 | 8087 | UTR 3 | XP_016857245.1 | ||
XM_017001757.1 | 8087 | UTR 3 | XP_016857246.1 |