Product Details

SNP ID
rs16877433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:108206901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATATTCTAAGACTCATTCCAACAT[A/G]GCTGGTGTAATTTTGCAACAAGCAT
Phenotype
MIM: 602210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF3E PubMed Links

Gene Details

Gene
EIF3E
Gene Name
eukaryotic translation initiation factor 3 subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001568.2 Intron NP_001559.1
XM_017013398.1 Intron XP_016868887.1

View Full Product Details