Product Details

SNP ID
rs17046782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55232273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAAACACCGACTTCAGTGCTTA[A/G]AGATAGTAACTAGGGAATCTGGGAG
Phenotype
MIM: 603766 MIM: 191343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLHC1 PubMed Links

Gene Details

Gene
CLHC1
Gene Name
clathrin heavy chain linker domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135598.1 41 Intron NP_001129070.1
NM_152385.2 41 UTR 5 NP_689598.2
XM_005264136.2 41 UTR 5 XP_005264193.1
XM_006711937.3 41 Intron XP_006712000.2
XM_006711938.3 41 Intron XP_006712001.1
XM_006711939.3 41 Intron XP_006712002.1
XM_011532519.2 41 UTR 5 XP_011530821.1
XM_011532520.2 41 UTR 5 XP_011530822.1
XM_011532521.2 41 UTR 5 XP_011530823.1
XM_011532522.2 41 UTR 5 XP_011530824.1
XM_011532523.2 41 UTR 5 XP_011530825.1
XM_011532524.1 41 Intron XP_011530826.1
XM_011532525.2 41 Intron XP_011530827.1
XM_011532527.2 41 UTR 5 XP_011530829.1
XM_011532528.1 41 UTR 5 XP_011530830.1
XM_011532529.2 41 UTR 5 XP_011530831.1
XM_011532530.1 41 Intron XP_011530832.1
XM_011532534.1 41 UTR 5 XP_011530836.1
XM_011532535.2 41 Intron XP_011530837.1
XM_017003333.1 41 UTR 5 XP_016858822.1
XM_017003334.1 41 UTR 5 XP_016858823.1
XM_017003335.1 41 UTR 5 XP_016858824.1
XM_017003336.1 41 UTR 5 XP_016858825.1
XM_017003337.1 41 Intron XP_016858826.1
XM_017003338.1 41 UTR 5 XP_016858827.1
XM_017003339.1 41 UTR 5 XP_016858828.1
XM_017003340.1 41 UTR 5 XP_016858829.1
XM_017003341.1 41 UTR 5 XP_016858830.1
XM_017003342.1 41 Intron XP_016858831.1
XM_017003343.1 41 UTR 5 XP_016858832.1
XM_017003344.1 41 UTR 5 XP_016858833.1
XM_017003345.1 41 UTR 5 XP_016858834.1
XM_017003346.1 41 UTR 5 XP_016858835.1
XM_017003347.1 41 UTR 5 XP_016858836.1
XM_017003348.1 41 UTR 5 XP_016858837.1
XM_017003349.1 41 Intron XP_016858838.1
XM_017003350.1 41 UTR 5 XP_016858839.1
Gene
MTIF2
Gene Name
mitochondrial translational initiation factor 2
There are no transcripts associated with this gene.

Gene
RPS27A
Gene Name
ribosomal protein S27a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135592.2 41 Intron NP_001129064.1
NM_001177413.1 41 Intron NP_001170884.1
NM_002954.5 41 Intron NP_002945.1

View Full Product Details