Product Details
- SNP ID
-
rs8002471
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:108499304 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGACCTTGACTTTGAAGATTGTAAA[C/T]TTCTAAGGACTGGCGAGGTGTGCTC
- Phenotype
-
MIM: 615479
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYO16
PubMed Links
Gene Details
- Gene
- MYO16
- Gene Name
- myosin XVI
View Full Product Details