Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004540.3 | Intron | NP_004531.2 | |||
XM_011529575.2 | Intron | XP_011527877.1 | |||
XM_011529576.2 | Intron | XP_011527878.1 | |||
XM_011529579.2 | Intron | XP_011527881.1 | |||
XM_011529580.2 | Intron | XP_011527882.1 | |||
XM_011529581.2 | Intron | XP_011527883.1 | |||
XM_011529582.2 | Intron | XP_011527884.1 | |||
XM_011529585.2 | Intron | XP_011527887.1 | |||
XM_017028353.1 | Intron | XP_016883842.1 | |||
XM_017028354.1 | Intron | XP_016883843.1 | |||
XM_017028355.1 | Intron | XP_016883844.1 | |||
XM_017028356.1 | Intron | XP_016883845.1 | |||
XM_017028357.1 | Intron | XP_016883846.1 | |||
XM_017028358.1 | Intron | XP_016883847.1 |