Product Details

SNP ID
rs944325
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:152797116 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTCTGCCTGCAAGGGAAGAACT[A/G]GAGCTGTGGACATGGGACTAAAGAC
Phenotype
MIM: 612605 MIM: 612606 MIM: 612607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCE1C PubMed Links

Gene Details

Gene
LCE1C
Gene Name
late cornified envelope 1C
There are no transcripts associated with this gene.

Gene
LCE1D
Gene Name
late cornified envelope 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178352.2 Intron NP_848129.1
Gene
LCE1E
Gene Name
late cornified envelope 1E
There are no transcripts associated with this gene.

View Full Product Details