Product Details
- SNP ID
-
rs7538207
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:21984181 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGACCCCTGACTCGGTGCTTTTTAT[C/T]GCTGCAGCCAACGGGCCACTCCCAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CELA3B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114040584] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CELA3B
- Gene Name
- chymotrypsin like elastase family member 3B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007352.3 |
|
Intron |
|
|
NP_031378.1 |
View Full Product Details