Product Details

SNP ID
rs4988288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135839638 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGTATTCACTCTTGTTTTTAATG[C/T]TTGCCATTTATTTTTAAAATACTTA
Phenotype
MIM: 603202 MIM: 601806
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCT PubMed Links

Gene Details

Gene
LCT
Gene Name
lactase
There are no transcripts associated with this gene.

Gene
MCM6
Gene Name
minichromosome maintenance complex component 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005915.5 3775 UTR 3 NP_005906.2

View Full Product Details