Product Details

SNP ID
rs11542580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69365108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATGATGGATTAATCAGAACATT[A/C]TGAACTGCTGAGGTTGCAGGAATTG
Phenotype
MIM: 603139 MIM: 600822
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AK6 PubMed Links

Gene Details

Gene
AK6
Gene Name
adenylate kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015891.1 1097 Intron NP_001015891.1
NM_016283.4 1097 Intron NP_057367.1
Gene
RAD17
Gene Name
RAD17 checkpoint clamp loader component
There are no transcripts associated with this gene.

Gene
TAF9
Gene Name
TATA-box binding protein associated factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015892.1 1097 Missense Mutation CAG,CAT Q210H NP_001015892.1
NM_003187.4 1097 Intron NP_003178.1

View Full Product Details