Product Details

SNP ID
rs7795717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152055661 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAATTAAATATTTCTCAAAAAC[G/T]AGAATTGGTGCTTATTACTATCTTA
Phenotype
MIM: 615130
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GALNT11 PubMed Links

Gene Details

Gene
GALNT11
Gene Name
polypeptide N-acetylgalactosaminyltransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304514.1 Intron NP_001291443.1
NM_022087.3 Intron NP_071370.2
XM_006716082.2 Intron XP_006716145.1
XM_006716083.2 Intron XP_006716146.1
XM_006716084.2 Intron XP_006716147.1
XM_017012499.1 Intron XP_016867988.1
XM_017012500.1 Intron XP_016867989.1

View Full Product Details