Product Details

SNP ID
rs4987862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63125074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTAGACATGTGTTGGGATTGCCC[C/T]GATTATTTACATTTAATCTTGATTA
Phenotype
MIM: 151430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL2 PubMed Links

Gene Details

Gene
BCL2
Gene Name
BCL2, apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000633.2 4764 UTR 3 NP_000624.2
NM_000657.2 4764 Intron NP_000648.2
XM_011526135.2 4764 Intron XP_011524437.1
XM_017025917.1 4764 Intron XP_016881406.1

View Full Product Details