Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_198317.2 | 408 | Intron | NP_938073.1 | ||
XM_006710600.3 | 408 | Intron | XP_006710663.1 | ||
XM_006710601.3 | 408 | Intron | XP_006710664.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001160184.1 | 408 | Intron | NP_001153656.1 | ||
NM_032129.2 | 408 | Intron | NP_115505.2 | ||
XM_006710944.3 | 408 | Missense Mutation | CCG,CGG | P7R | XP_006711007.2 |
XM_011542248.2 | 408 | Missense Mutation | CCG,CGG | P7R | XP_011540550.2 |
XM_017002474.1 | 408 | Missense Mutation | CCG,CGG | P7R | XP_016857963.1 |
XM_017002475.1 | 408 | Missense Mutation | CCG,CGG | P7R | XP_016857964.1 |
XM_017002476.1 | 408 | Missense Mutation | CCG,CGG | P7R | XP_016857965.1 |
XM_017002477.1 | 408 | Missense Mutation | CCG,CGG | P7R | XP_016857966.1 |
XM_017002478.1 | 408 | Missense Mutation | CCG,CGG | P7R | XP_016857967.1 |
XM_017002479.1 | 408 | Missense Mutation | CCG,CGG | P7R | XP_016857968.1 |