Product Details

SNP ID
rs28363531
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:171100164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAGACGTTGCTGAAGATAGAAC[A/G]CACTGCTGATATGGCTATTGAGAGG
Phenotype
MIM: 136132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FMO3 PubMed Links

Gene Details

Gene
FMO3
Gene Name
flavin containing monooxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002294.2 Intron NP_001002294.1
NM_001319173.1 Intron NP_001306102.1
NM_001319174.1 Intron NP_001306103.1
NM_006894.5 Intron NP_008825.4
XM_011509345.2 Intron XP_011507647.1
Gene
MIR1295A
Gene Name
microRNA 1295a
There are no transcripts associated with this gene.

Gene
MIR1295B
Gene Name
microRNA 1295b
There are no transcripts associated with this gene.

View Full Product Details