Product Details
- SNP ID
-
rs28575430
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:198811262 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGAGAGTCAGGAAATTCATAGC[A/G]ATAAGGACATGGAAGAAGAGCATAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR181A1HG
PubMed Links
Gene Details
- Gene
- MIR181A1HG
- Gene Name
- MIR181A1 host gene
There are no transcripts associated with this gene.
View Full Product Details