Product Details

SNP ID
rs28722116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112090688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTAATTAGTGGTTTTTTTCAAC[A/C]CACCAGAGTCCTAATTTGTGATCTC
Phenotype
MIM: 611956
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C3orf52 PubMed Links

Gene Details

Gene
C3orf52
Gene Name
chromosome 3 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171747.1 Intron NP_001165218.1
NM_024616.2 Intron NP_078892.2
XM_011513133.2 Intron XP_011511435.1
XM_017007181.1 Intron XP_016862670.1
Gene
TMPRSS7
Gene Name
transmembrane protease, serine 7
There are no transcripts associated with this gene.

View Full Product Details