Product Details

SNP ID
rs9273916
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32661969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAACTCAATATCCCCTTACGC[C/G]ACTCCACGGTGATGGGGCTCTGGAG
Phenotype
MIM: 146880 MIM: 604305
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
HLA-DQA1 PubMed Links

Gene Details

Gene
HLA-DQA1
Gene Name
major histocompatibility complex, class II, DQ alpha 1
There are no transcripts associated with this gene.

Gene
HLA-DQB1
Gene Name
major histocompatibility complex, class II, DQ beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243961.1 741 Missense Mutation TCG,TGG S220W NP_001230890.1
NM_001243962.1 741 Missense Mutation TCG,TGG S220W NP_001230891.1
NM_002123.4 741 Missense Mutation TCG,TGG S220W NP_002114.3
Gene
HLA-DQB1-AS1
Gene Name
HLA-DQB1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details