Product Details

SNP ID
rs28661532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64983676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTACACCTGCATGCTGAGAATA[C/T]GCTTTTAAAAGTGTCAGCACCACCT
Phenotype
MIM: 131170 MIM: 194624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERV3-1 PubMed Links

Gene Details

Gene
ERV3-1
Gene Name
endogenous retrovirus group 3 member 1
There are no transcripts associated with this gene.

Gene
ZNF117
Gene Name
zinc finger protein 117
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015852.3 Intron NP_056936.2

View Full Product Details