Product Details

SNP ID
rs28652910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41394844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCTCATTTTTATAAACAATGAA[A/G]ATTTATGCTGTTACCTTGACCTCCT
Phenotype
MIM: 606934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFAF1 PubMed Links

Gene Details

Gene
NDUFAF1
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016013.3 Intron NP_057097.2
XM_006720555.2 Intron XP_006720618.1
XM_011521658.1 Intron XP_011519960.1
XM_011521659.2 Intron XP_011519961.1
XM_017022293.1 Intron XP_016877782.1

View Full Product Details