Product Details

SNP ID
rs28528124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23657805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATATCCTCTAGGAAAAATGTCT[A/G]AGTTTCTGTGCTTAGCATACATAAT
Phenotype
MIM: 612962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCTN5 PubMed Links

Gene Details

Gene
DCTN5
Gene Name
dynactin subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199011.1 Intron NP_001185940.1
NM_001199743.1 Intron NP_001186672.1
NM_032486.3 Intron NP_115875.1

View Full Product Details