Product Details

SNP ID
rs35880761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74874852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTTTACATATTGTCTATGGCT[A/G]TATTTGTGCTTTGATGGCACAATTG
Phenotype
MIM: 103270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FADS6 PubMed Links

Gene Details

Gene
FADS6
Gene Name
fatty acid desaturase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178128.5 4012 Intron NP_835229.3
XM_005257224.4 4012 UTR 3 XP_005257281.2
XM_017024458.1 4012 UTR 3 XP_016879947.1
Gene
FDXR
Gene Name
ferredoxin reductase
There are no transcripts associated with this gene.

View Full Product Details