Product Details

SNP ID
rs35970532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35271201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCACCCTCACAGGCTCAGCCAGC[C/T]CTGGAGTGTGGAGTGACAGAGAGAG
Phenotype
MIM: 616582 MIM: 600390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSR PubMed Links

Gene Details

Gene
LSR
Gene Name
lipolysis stimulated lipoprotein receptor
There are no transcripts associated with this gene.

Gene
USF2
Gene Name
upstream transcription factor 2, c-fos interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321150.1 Intron NP_001308079.1
NM_003367.3 Intron NP_003358.1
NM_207291.2 Intron NP_997174.1
XM_005259197.3 Intron XP_005259254.1
XM_011527260.2 Intron XP_011525562.1
XM_011527261.2 Intron XP_011525563.1
XM_017027199.1 Intron XP_016882688.1

View Full Product Details