Product Details
- SNP ID
-
rs28687244
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23605228 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGTCTGTCTGTCTCTCTCTGTCT[C/G]TCTCTCTCTCTCCCTCTGTGTGTCC
- Phenotype
-
MIM: 616543
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CST9
PubMed Links
Gene Details
- Gene
- CST9
- Gene Name
- cystatin 9
View Full Product Details