Product Details

SNP ID
rs17885940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:31659731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGCGTCTGGGGTTTCCGTTGC[A/G]GTCCTCGGAACCAGGACCTCGGCGT
Phenotype
MIM: 147450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724449 PubMed Links

Gene Details

Gene
LOC102724449
Gene Name
uncharacterized LOC102724449
There are no transcripts associated with this gene.

Gene
SOD1
Gene Name
superoxide dismutase 1, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000454.4 110 UTR 5 NP_000445.1

View Full Product Details