Product Details

SNP ID
rs28434564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22515973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGCCTCTAAATTTTGCACTCTA[G/T]ATGCCTCACTCACCTCCTCCTCATC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF280A PubMed Links

Gene Details

Gene
ZNF280A
Gene Name
zinc finger protein 280A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080740.4 Intron NP_542778.2
Gene
ZNF280B
Gene Name
zinc finger protein 280B
There are no transcripts associated with this gene.

View Full Product Details