Product Details
- SNP ID
-
rs2229013
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:8535755 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACCGGTGATGTTCACATCCTGGA[T/C]GATGAATGAAGCAGAAAGGTTCTCA
- Phenotype
-
MIM: 300836
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANOS1
PubMed Links
Gene Details
- Gene
- ANOS1
- Gene Name
- anosmin 1
View Full Product Details