Product Details

SNP ID
rs28373336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:104472626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATATAGGAAATGCAGCCTGAAA[A/C]ACGTATAGGAAGCAGAATAAAGGTA
Phenotype
MIM: 611645
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CXXC4 PubMed Links

Gene Details

Gene
CXXC4
Gene Name
CXXC finger protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025212.3 Intron NP_079488.2
XM_011532284.2 Intron XP_011530586.1
XM_017008652.1 Intron XP_016864141.1

View Full Product Details