Product Details

SNP ID
rs28790702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49957258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTGCTGGGATTACAGATGTTA[A/G]CCACTGGCCTGTTGTGAAAATGTTT
Phenotype
MIM: 606886
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM6 PubMed Links

Gene Details

Gene
RBM6
Gene Name
RNA binding motif protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167582.1 Intron NP_001161054.1
NM_005777.2 Intron NP_005768.1
XM_005264784.1 Intron XP_005264841.1
XM_005264785.1 Intron XP_005264842.1
XM_005264786.1 Intron XP_005264843.1
XM_005264787.2 Intron XP_005264844.1
XM_006712916.1 Intron XP_006712979.1
XM_017005496.1 Intron XP_016860985.1
XM_017005497.1 Intron XP_016860986.1
XM_017005498.1 Intron XP_016860987.1
XM_017005499.1 Intron XP_016860988.1
XM_017005500.1 Intron XP_016860989.1
XM_017005501.1 Intron XP_016860990.1
XM_017005502.1 Intron XP_016860991.1

View Full Product Details