Product Details

SNP ID
rs35843847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:89956202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAGGGGGCGCGCACGGGCGCT[C/G]CGGAGACCTGTGTCCTGCCGGGGCG
Phenotype
MIM: 607198
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB11 PubMed Links

Gene Details

Gene
EFCAB11
Gene Name
EF-hand calcium binding domain 11
There are no transcripts associated with this gene.

Gene
TDP1
Gene Name
tyrosyl-DNA phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008744.1 214 Intron NP_001008744.1
NM_018319.3 214 Intron NP_060789.2
XM_005267847.2 214 Intron XP_005267904.1
XM_005267848.2 214 UTR 5 XP_005267905.1
XM_006720197.3 214 UTR 5 XP_006720260.1
XM_006720198.3 214 Intron XP_006720261.1
XM_006720199.1 214 Intron XP_006720262.1
XM_011536942.2 214 Intron XP_011535244.1
XM_011536943.2 214 Intron XP_011535245.1
XM_011536944.1 214 Intron XP_011535246.1
XM_017021439.1 214 Intron XP_016876928.1
XM_017021440.1 214 Intron XP_016876929.1
XM_017021441.1 214 UTR 5 XP_016876930.1
XM_017021442.1 214 Intron XP_016876931.1
XM_017021443.1 214 Intron XP_016876932.1
XM_017021444.1 214 Intron XP_016876933.1
XM_017021445.1 214 Intron XP_016876934.1
XM_017021446.1 214 UTR 5 XP_016876935.1

View Full Product Details