Product Details

SNP ID
rs35947490
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:52606277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCCCAGGTCTCCCTTTGTACCC[A/C]CTCTTATCAGTTCTCAGTATGTGCT
Phenotype
MIM: 615784
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPX7 PubMed Links

Gene Details

Gene
GPX7
Gene Name
glutathione peroxidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015696.4 Intron NP_056511.2

View Full Product Details