Product Details

SNP ID
rs28383597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3002060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTTAACCGCTAGGGGGATGGAG[A/G]CAGAAGTCACAAACTCACATGCCAC
Phenotype
MIM: 160998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NQO2 PubMed Links

Gene Details

Gene
NQO2
Gene Name
NAD(P)H quinone dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000904.4 281 Intron NP_000895.2
NM_001290221.1 281 UTR 5 NP_001277150.1
NM_001290222.1 281 Intron NP_001277151.1
NM_001318940.1 281 Intron NP_001305869.1

View Full Product Details