Product Details

SNP ID
rs72704879
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:153040515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCAAGACTACTTCGTTTCTCC[C/T]TTCCACTTTAGCAAATTGCTTCATT
Phenotype
MIM: 182266
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPRR1B PubMed Links

Gene Details

Gene
SPRR1B
Gene Name
small proline rich protein 1B
There are no transcripts associated with this gene.

Gene
SPRR2D
Gene Name
small proline rich protein 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006945.4 Intron NP_008876.3
XM_017002175.1 Intron XP_016857664.1

View Full Product Details