Product Details

SNP ID
rs61735573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110476429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGCAAAAATGAACTTTGCGGA[A/G]AGAGAGGGCTCTAAGAGATACTGCA
Phenotype
MIM: 138297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENPEP PubMed Links

Gene Details

Gene
ENPEP
Gene Name
glutamyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001977.3 357 Silent Mutation GAA,GAG E5E NP_001968.3
XM_017007877.1 357 Intron XP_016863366.1

View Full Product Details