Product Details

SNP ID
rs61739330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74415827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCGTCCTCACCCCATGTGGGG[G/T]CCTCAGCTACTGCAGATTCTCCCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145054.1 420 Missense Mutation GAC,GCC D120A NP_001138526.1
NM_001316764.1 420 Missense Mutation GAC,GCC D117A NP_001303693.1
NM_001316765.1 420 Missense Mutation GAC,GCC D68A NP_001303694.1
NM_001316766.1 420 Missense Mutation GAC,GCC D22A NP_001303695.1
Gene
WDR54
Gene Name
WD repeat domain 54
There are no transcripts associated with this gene.

View Full Product Details