Product Details

SNP ID
rs41270569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:81746866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATTTCACTGTGTGTTCAACTAC[A/T]TATGCACAAGACTATAGTACACACA
Phenotype
MIM: 611357
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM46A PubMed Links

Gene Details

Gene
FAM46A
Gene Name
family with sequence similarity 46 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017633.2 4476 UTR 3 NP_060103.2

View Full Product Details