Product Details

SNP ID
rs45480798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33111276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAAAAAAAAAAAAAAAA[A/C]AACAACAAGAAAAGGTAGAGTTTGG
Phenotype
MIM: 137060
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
B4GALT1 PubMed Links

Gene Details

Gene
B4GALT1
Gene Name
beta-1,4-galactosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001497.3 3474 UTR 3 NP_001488.2
XM_005251440.4 3474 UTR 3 XP_005251497.1

View Full Product Details