Product Details

SNP ID
rs41292396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28986216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCCAGACCTGCTCCCCATCC[C/T]GTGAGTAGCTGCTCAGCCCCTGCCC
Phenotype
MIM: 602354 MIM: 612583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAT PubMed Links

Gene Details

Gene
LAT
Gene Name
linker for activation of T-cells
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014987.1 597 Missense Mutation CCA,CTA P82L NP_001014987.1
NM_001014988.1 597 Missense Mutation CCA,CTA P82L NP_001014988.1
NM_001014989.1 597 Missense Mutation CCA,CTA P118L NP_001014989.2
NM_014387.3 597 Missense Mutation CCA,CTA P82L NP_055202.1
Gene
SPNS1
Gene Name
sphingolipid transporter 1 (putative)
There are no transcripts associated with this gene.

View Full Product Details