Product Details
- SNP ID
-
rs41310051
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:76385716 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAGATGTAAAAGGACAATAACAT[A/G]TCAAAGAACTTTCACACACCTAAAG
- Phenotype
-
MIM: 613010
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RFK
PubMed Links
Gene Details
- Gene
- RFK
- Gene Name
- riboflavin kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018339.5 |
2508 |
UTR 3 |
|
|
NP_060809.3 |
View Full Product Details