Product Details

SNP ID
rs62278666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:157545495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACTATGATCCCTTCTTGACCGC[C/T]TGGGAAAGACTCAAACTCTCTGAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PQLC2L PubMed Links

Gene Details

Gene
PQLC2L
Gene Name
PQ loop repeat containing 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099777.3 Intron NP_001093247.1
NM_001130001.2 Intron NP_001123473.1
NM_001130002.2 Intron NP_001123474.1
NM_001243729.1 Intron NP_001230658.1
XM_017005754.1 Intron XP_016861243.1

View Full Product Details